Learn More
Medchemexpress LLC 8-bromo-cAMP sodium salt | 76939-46-3 | MFCD00149373 | 99.9% | 430.08 g/mol | C10H10BrN5NaO6P | 50MG
SDP
Catalog No. 5000236339
Encompass_Preferred
8-Bromo-cAMP sodium salt is a cell-permeable cyclic AMP analog that activates cyclic AMP-dependent protein kinase (PKA). Supplied as a white to off-white solid with high purity, it is used to elevate intracellular cAMP for in vitro and in vivo studies of cAMP-mediated signaling, apoptosis, and related pathways. Manufacturer provides solubility data, formulation protocols, and storage recommendations for research use.
  • Cell-permeable cyclic AMP analog.
  • Activates cyclic AMP-dependent protein kinase (PKA).
  • High purity suitable for research applications.
  • Soluble in DMSO and water; manufacturer provides formulation protocols.
  • Available in multiple pack sizes with defined storage guidance.
Catalog No. 50-002-36339 Supplier Medchemexpress LLC Supplier No. HY1230650MG
8-Bromo-cAMP sodium salt is a cell-permeable cyclic AMP analog that activates cyclic AMP-dependent protein kinase (PKA). Supplied as a white to off-white solid with high purity, it is used to elevate intracellular cAMP for in vitro and in vivo studies of cAMP-mediated signaling, apoptosis, and related pathways. Manufacturer provides solubility data, formulation protocols, and storage recommendations for research use.
  • Cell-permeable cyclic AMP analog.
  • Activates cyclic AMP-dependent protein kinase (PKA).
  • High purity suitable for research applications.
  • Soluble in DMSO and water; manufacturer provides formulation protocols.
  • Available in multiple pack sizes with defined storage guidance.