Learn More
Enzo Life Sciences 8-Bromo-cGMP (50mg). CAS: 51116-01-9
SDP
Catalog No. 502010449
Encompass_Preferred
A cell-permeable cGMP analog which is more resistant to phosphodiesterases than cGMP and which preferentially activates cGMP-dependent protein kinase. It inhibits thrombin stimulated arachidonic acid release in human platelets. Purity: ≥96% (HPLC). Solubility: Soluble in water (25mg/ml) or aqueous buffers. Sparingly soluble in methanol (4mM). Insoluble in DMSO, dimethyl formamide and 96% ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0449 Supplier Enzo Life Sciences Supplier No. BMLCN2050050
A cell-permeable cGMP analog which is more resistant to phosphodiesterases than cGMP and which preferentially activates cGMP-dependent protein kinase. It inhibits thrombin stimulated arachidonic acid release in human platelets. Purity: ≥96% (HPLC). Solubility: Soluble in water (25mg/ml) or aqueous buffers. Sparingly soluble in methanol (4mM). Insoluble in DMSO, dimethyl formamide and 96% ethanol. Long Term Storage: -20°C.