Learn More
Medchemexpress LLC HY-101379A 10mg Medchemexpress, 8-Bromo-cGMP (sodium) CAS:51116-01-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101379A10MG
Encompass_Preferred
Medchemexpress, HY-101379A 10mg 8-Bromo-cGMP (sodium) CAS:51116-01-9 8-Bromo-cGMP sodium, a membrane-permeable analogue of cGMP, is a PKG (protein kinase G) activator and significantly inhibits Ca2+ macroscopic currents and impairs insulin release stimulated with high K+. 8-Bromo-cGMP sodium has antinociceptive effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7837