Learn More
Medchemexpress LLC 8-bromo-cGMP sodium | 51116-01-9 | 99.9% | 446.08 | C10H10BrN5NaO7P | 100 MG
SDP
Catalog No. 5000213222
Encompass_Preferred
8-Bromo-cGMP sodium is a membrane-permeable cyclic GMP analogue that activates protein kinase G (PKG). Supplied as the sodium salt in solid form at high purity, it is used as a research reagent to probe cGMP signaling pathways, modulate ion channel activity, and study vascular and neuronal responses. Store sealed at -20°C; in solvent, store at -80°C for longer-term stability.
  • Membrane-permeable analogue of cGMP that activates protein kinase G (PKG).
  • High purity (99.9%) suitable for biochemical and cellular assays.
  • Off-white to light yellow solid for straightforward handling.
  • Available in lab-scale quantities, including 100 MG.
  • Stable when stored sealed at -20°C; in solvent, store at -80°C for up to 6 months.
  • Used to study cGMP signaling, ion channel modulation, and vascular or neuronal pharmacology.
Catalog No. 50-002-13222 Supplier Medchemexpress LLC Supplier No. HY101379A100MG
8-Bromo-cGMP sodium is a membrane-permeable cyclic GMP analogue that activates protein kinase G (PKG). Supplied as the sodium salt in solid form at high purity, it is used as a research reagent to probe cGMP signaling pathways, modulate ion channel activity, and study vascular and neuronal responses. Store sealed at -20°C; in solvent, store at -80°C for longer-term stability.
  • Membrane-permeable analogue of cGMP that activates protein kinase G (PKG).
  • High purity (99.9%) suitable for biochemical and cellular assays.
  • Off-white to light yellow solid for straightforward handling.
  • Available in lab-scale quantities, including 100 MG.
  • Stable when stored sealed at -20°C; in solvent, store at -80°C for up to 6 months.
  • Used to study cGMP signaling, ion channel modulation, and vascular or neuronal pharmacology.