Learn More
Apexbio Technology LLC 8-Bromo-cGMP, sodium salt 51116-01-9 50mg
SDP
Catalog No. 5000092254
Encompass_Preferred
8-Bromo-cGMP sodium salt (CAS 51116-01-9) is a membrane-permeable analog of cyclic GMP that functions as an activator of protein kinase G (PKG) By mimicking endogenous cGMP it modulates intracellular signaling pathways notably influencing Ca2 channel activity Studies indicate that 8-Bromo-cGMP sodium salt can suppress macroscopic ionic currents and reduce insulin secretion under high K stimulation Due to its role in signal transduction and vascular function this compound is frequently employed in research addressing nociceptive modulation and vascular biology
Catalog No. 50-000-92254 Supplier Apexbio Technology LLC Supplier No. B659450
8-Bromo-cGMP sodium salt (CAS 51116-01-9) is a membrane-permeable analog of cyclic GMP that functions as an activator of protein kinase G (PKG) By mimicking endogenous cGMP it modulates intracellular signaling pathways notably influencing Ca2 channel activity Studies indicate that 8-Bromo-cGMP sodium salt can suppress macroscopic ionic currents and reduce insulin secretion under high K stimulation Due to its role in signal transduction and vascular function this compound is frequently employed in research addressing nociceptive modulation and vascular biology