Learn More
Cayman Chemical 8-bromo Cyclic AMP, Purity: ≥95%, CAS: 23583-48-4, 100 mg
SDP
Catalog No. 502964892
Encompass_Preferred
A PKA activator (Ka = 0.05 µM); selective for PKA over PKG (Ka = 5.8 µM); more resistant to hydrolysis by PDEs than cAMP; inhibits the proliferation of HL-60 cells (IC50 = 18 µM after six days); induces relaxation of phenylephrine-precontracted isolated rabbit carotid or renal arterial rings at 10-300 µM; intradermal administration prevents SLIGRL-NH2-induced scratching and increases in cutaneous LTB4 production in mice at 10 nmol/site
Catalog No. 50-296-4892 Supplier Cayman Chemical Supplier No. 18141100 MG
A PKA activator (Ka = 0.05 µM); selective for PKA over PKG (Ka = 5.8 µM); more resistant to hydrolysis by PDEs than cAMP; inhibits the proliferation of HL-60 cells (IC50 = 18 µM after six days); induces relaxation of phenylephrine-precontracted isolated rabbit carotid or renal arterial rings at 10-300 µM; intradermal administration prevents SLIGRL-NH2-induced scratching and increases in cutaneous LTB4 production in mice at 10 nmol/site