Learn More
Cayman Chemical 8-bromo NAD+ (sodium salt), Purity: ≥95%, CAS: 2022926-16-3, 7.4 mg
SDP
Catalog No. 502917126
Encompass_Preferred
A prodrug form of 8-bromo-cADPR; prevents fMLP-induced increases in intracellular calcium levels and chemotaxis in isolated mouse bone marrow-derived neutrophils at 1 mM; decreases LPS-induced nitrite production and TNF-α and IL-2 secretion in mouse primary microglial cells at 100 µM
Catalog No. 50-291-7126 Supplier Cayman Chemical Supplier No. 384757.4 MG
A prodrug form of 8-bromo-cADPR; prevents fMLP-induced increases in intracellular calcium levels and chemotaxis in isolated mouse bone marrow-derived neutrophils at 1 mM; decreases LPS-induced nitrite production and TNF-α and IL-2 secretion in mouse primary microglial cells at 100 µM