Learn More
Medchemexpress LLC 8-Bromoadenosine | 2946-39-6 | 99.73% | 346.14 | 5 G
SDP
Catalog No. 5000454371
Encompass_Preferred
8-Bromoadenosine is an adenosine analog. Adenosine analogs primarily function as smooth muscle vasodilators and have been shown to hinder cancer progression. It can quench the intrinsic fluorescence of human serum albumin (HSA) through a static quenching procedure. It can be utilized to synthesize adenosine phosphates, such as Acadesine, Clofarabine, Fludarabine phosphate, and Vidarabine.
  • Reduces the fluorescence emission intensity of HSA.
  • Binds to HSA with reduced stability as temperature increases, though the temperature effect is minimal.
  • Exhibits decreased binding constants with HSA in the presence of other ions.
Catalog No. 50-004-54371 Supplier Medchemexpress LLC Supplier No. HYW0131955G
8-Bromoadenosine is an adenosine analog. Adenosine analogs primarily function as smooth muscle vasodilators and have been shown to hinder cancer progression. It can quench the intrinsic fluorescence of human serum albumin (HSA) through a static quenching procedure. It can be utilized to synthesize adenosine phosphates, such as Acadesine, Clofarabine, Fludarabine phosphate, and Vidarabine.
  • Reduces the fluorescence emission intensity of HSA.
  • Binds to HSA with reduced stability as temperature increases, though the temperature effect is minimal.
  • Exhibits decreased binding constants with HSA in the presence of other ions.