Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-bromoadenosine 3',5'-cyclic monophosphate | 23583-48-4 | MFCD00075580 | 99.5% | 408.10 g/mol | C10H11BrN5O6P | 500MG
SDP

Catalog No. 5000238388
Encompass_Preferred

8-Bromo-cAMP (8-Br-cAMP) is a membrane-permeable brominated cyclic AMP analog used to elevate intracellular cAMP and activate cyclic AMP-dependent protein kinase (PKA) in cell signaling research. It is applied in cell-based assays to probe cAMP/PKA-mediated pathways and has reported anti-proliferative and pro-apoptotic effects in some cancer cell models.

  • Activator of cyclic AMP-dependent protein kinase (PKA).
  • Membrane-permeable cAMP analog that raises intracellular cAMP.
  • Useful for studying cAMP/PKA signaling pathways in cell-based assays.
  • Reported to induce anti-proliferative and pro-apoptotic effects in cancer cell models.
  • High reported purity suitable for research applications.
  • Chemical formula C10H11BrN5O6P; molecular weight 408.10 g/mol.

Catalog No. 50-002-38388 Supplier Medchemexpress LLC Supplier No. HY12306A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Bromo-cAMP (8-Br-cAMP) is a membrane-permeable brominated cyclic AMP analog used to elevate intracellular cAMP and activate cyclic AMP-dependent protein kinase (PKA) in cell signaling research. It is applied in cell-based assays to probe cAMP/PKA-mediated pathways and has reported anti-proliferative and pro-apoptotic effects in some cancer cell models.

  • Activator of cyclic AMP-dependent protein kinase (PKA).
  • Membrane-permeable cAMP analog that raises intracellular cAMP.
  • Useful for studying cAMP/PKA signaling pathways in cell-based assays.
  • Reported to induce anti-proliferative and pro-apoptotic effects in cancer cell models.
  • High reported purity suitable for research applications.
  • Chemical formula C10H11BrN5O6P; molecular weight 408.10 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.