Learn More
Medchemexpress LLC 8-bromoadenosine 3',5'-cyclic monophosphate | 23583-48-4 | MFCD00075580 | 99.5% | 408.10 g/mol | C10H11BrN5O6P | 5MG
SDP
Catalog No. 5000238389
Encompass_Preferred
8-Bromo-cAMP is a cell-permeable cyclic AMP analog used to activate cyclic AMP-dependent protein kinase (PKA) in biochemical and cell signaling research. It is more resistant to degradation by cyclic AMP phosphodiesterases than cAMP and has been used in studies reporting anti-proliferative and pro-apoptotic effects in cellular models.
  • Cell-permeable cyclic AMP analog suitable for cell-based assays.
  • Activates cyclic AMP-dependent protein kinase (PKA).
  • Resistant to phosphodiesterase degradation for prolonged activity.
  • High purity (≈99.5%) appropriate for research use.
  • Solid, white to light yellow appearance for easy handling.
  • Powder stability: -20°C for long-term storage, 4°C for shorter-term storage.
  • Soluble in 1 M NaOH at high concentration with warming and sonication.
Catalog No. 50-002-38389 Supplier Medchemexpress LLC Supplier No. HY12306A5MG
8-Bromo-cAMP is a cell-permeable cyclic AMP analog used to activate cyclic AMP-dependent protein kinase (PKA) in biochemical and cell signaling research. It is more resistant to degradation by cyclic AMP phosphodiesterases than cAMP and has been used in studies reporting anti-proliferative and pro-apoptotic effects in cellular models.
  • Cell-permeable cyclic AMP analog suitable for cell-based assays.
  • Activates cyclic AMP-dependent protein kinase (PKA).
  • Resistant to phosphodiesterase degradation for prolonged activity.
  • High purity (≈99.5%) appropriate for research use.
  • Solid, white to light yellow appearance for easy handling.
  • Powder stability: -20°C for long-term storage, 4°C for shorter-term storage.
  • Soluble in 1 M NaOH at high concentration with warming and sonication.