Learn More
Medchemexpress LLC 8-bromoadenosine 5'-monophosphate | 23567-96-6 | MFCD00056945 | 98.8% | C10H13BrN5O7P | 10MG
SDP
Supplier:  Medchemexpress LLC HY13426610MG
Encompass_Preferred
8-Bromo-AMP (8-Bromoadenosine 5'-monophosphate) is a membrane-permeable cAMP analogue used in biochemical and pharmacological research. It is reported to improve cardiac recovery after ischemia-reperfusion by increasing ATP, ADP, and total adenine nucleotide levels. The compound is provided as a high-purity, water-soluble solid suitable for laboratory assays.
  • Membrane-permeable cAMP analogue suitable for cell-based studies.
  • Reported to support cardiac recovery after ischemia-reperfusion by elevating adenine nucleotides.
  • High purity (98.8%) suitable for research applications.
  • Water soluble (≈125 mg/mL; ultrasonic assistance may be required).
  • White to off-white solid, easy to handle and store.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-62053