Learn More
Cayman Chemical 8-Bromoadenosine 5′-triphosphate (sodium salt hydrate), Purity: ≥95%, 10 mg
SDP
Catalog No. 502978915
Encompass_Preferred
An ATP analog and P2X receptor agonist; induces contraction of isolated guinea pig bladder strips with a potency that is 0.19-fold that of ATP; has been used in the determiniation of the nucleotide substrate specificity of yeast poly(A) polymerase; cytotoxic to multiple myeloma cells (IC50 = 23.1 μM)
Catalog No. 50-297-8915 Supplier Cayman Chemical Supplier No. 3057310 MG
An ATP analog and P2X receptor agonist; induces contraction of isolated guinea pig bladder strips with a potency that is 0.19-fold that of ATP; has been used in the determiniation of the nucleotide substrate specificity of yeast poly(A) polymerase; cytotoxic to multiple myeloma cells (IC50 = 23.1 μM)