Learn More
Medchemexpress LLC 8-bromoguanosine | 4016-63-1 | MFCD00037985 | 99.6% | 362.14 g/mol | C10H12BrN5O5 | 25 G
SDP
Catalog No. 5000302831
Encompass_Preferred
8-Bromoguanosine is a brominated derivative of the purine nucleoside guanosine used as a research reagent to probe and modulate RNA structure and function. It reduces conformational heterogeneity in RNA and is employed in biochemical and biophysical experiments requiring a high-purity nucleoside analog.
  • Reduces conformational heterogeneity in RNA.
  • Useful for biochemical and structural RNA studies.
  • High purity suitable for analytical and experimental applications.
  • Soluble in DMSO; can be formulated for in vivo administration.
  • Available in multiple packaging sizes for laboratory use.
Catalog No. 50-003-02831 Supplier Medchemexpress LLC Supplier No. 25GHYW019033
8-Bromoguanosine is a brominated derivative of the purine nucleoside guanosine used as a research reagent to probe and modulate RNA structure and function. It reduces conformational heterogeneity in RNA and is employed in biochemical and biophysical experiments requiring a high-purity nucleoside analog.
  • Reduces conformational heterogeneity in RNA.
  • Useful for biochemical and structural RNA studies.
  • High purity suitable for analytical and experimental applications.
  • Soluble in DMSO; can be formulated for in vivo administration.
  • Available in multiple packaging sizes for laboratory use.