Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromoguanosine hydrate, 97%

Catalog No. AAL0299203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0299203 1 g
AAL0299206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0299203 Supplier Thermo Scientific Chemicals Supplier No. L0299203
Add to Cart

CAS: 332359-99-6 | C10H12BrN5O5 | 362.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332359-99-6
Molecular Formula C10H12BrN5O5
Molecular Weight (g/mol) 362.14
MDL Number MFCD00150531
InChI Key ASUCSHXLTWZYBA-UMMCILCDSA-N
Synonym 8-bromoguanosine hydrate, 8-bromoguanosine dihydrate, 2r,3r,4s,5r-2-2-amino-8-bromo-6-hydroxypurin-9-yl-5-hydroxymethyl oxolane-3,4-diol dihydrate, 3s,2r,4r,5r-5-2-amino-8-bromo-6-hydroxypurin-9-yl-2-hydroxymethyl oxolane-3,4-diol, hydrate, hydrate, 2-amino-8-bromo-9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one dihydrate
PubChem CID 18530343
IUPAC Name 2-amino-8-bromo-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one;dihydrate
SMILES NC1=NC2=C(N=C(Br)N2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 578725
Optical Rotation −30° (c=2 in 1:1 DMSO/Water)
Formula Weight 362.14 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 8-Bromoguanosine hydrate
EINECSNumber 223-677-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.