Learn More
Cayman Chemical 8-BromogunosIn-5-O-trIphos 3mg
SDP
Catalog No. 502918783
Encompass_Preferred
A GTP derivative and bacterial GTPase inhibitor; inhibits E. coli FtsZ (Ki = 31.8 µM) but promotes assembly of porcine brain microtubules in a cell-free assay at 500 µM
Catalog No. 50-291-8783 Supplier Cayman Chemical Supplier No. 385513 MG
A GTP derivative and bacterial GTPase inhibitor; inhibits E. coli FtsZ (Ki = 31.8 µM) but promotes assembly of porcine brain microtubules in a cell-free assay at 500 µM