Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-Bromoisoquinoline | 63927-22-0 | 99.51% | C9H6BrN | 5 G
SDP

Catalog No. 5000434273
Encompass_Preferred

8-Bromoisoquinoline is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is for research use only.

  • Purity: 99.51%
  • Molecular weight: 208.05
  • Formula: C9H6BrN
  • CAS No.: 63927-22-0
  • Appearance: Solid, off-white to yellow color

Catalog No. 50-004-34273 Supplier Medchemexpress LLC Supplier No. HY690895G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Bromoisoquinoline is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is for research use only.

  • Purity: 99.51%
  • Molecular weight: 208.05
  • Formula: C9H6BrN
  • CAS No.: 63927-22-0
  • Appearance: Solid, off-white to yellow color

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.