Learn More
Medchemexpress LLC 8-bromooct-1-yne | 81216-13-9 | MFCD20621358 | 189.09 | C8H13Br | 500 MG
SDP
Catalog No. 5000284586
Encompass_Preferred
8-Bromooct-1-yne is an organobromide reagent containing a terminal alkyne and a bromide leaving group. It is used as a synthetic building block in organic chemistry and for bioconjugation workflows such as copper-catalyzed azide-alkyne cycloaddition (click chemistry). Store under nitrogen at 4°C; in solution, keep at -80°C (up to 6 months) or -20°C (up to 1 month). CAS: 81216-13-9; formula: C8H13Br; MW: 189.09.
  • Terminal alkyne for conjugation and cross-coupling reactions.
  • Bromide leaving group enables nucleophilic substitution.
  • Suitable for copper-catalyzed click chemistry with azide partners.
  • Supplied in small-scale quantities for research applications.
  • Store under inert atmosphere to preserve stability.
Catalog No. 50-002-84586 Supplier Medchemexpress LLC Supplier No. HYW591970500MG
8-Bromooct-1-yne is an organobromide reagent containing a terminal alkyne and a bromide leaving group. It is used as a synthetic building block in organic chemistry and for bioconjugation workflows such as copper-catalyzed azide-alkyne cycloaddition (click chemistry). Store under nitrogen at 4°C; in solution, keep at -80°C (up to 6 months) or -20°C (up to 1 month). CAS: 81216-13-9; formula: C8H13Br; MW: 189.09.
  • Terminal alkyne for conjugation and cross-coupling reactions.
  • Bromide leaving group enables nucleophilic substitution.
  • Suitable for copper-catalyzed click chemistry with azide partners.
  • Supplied in small-scale quantities for research applications.
  • Store under inert atmosphere to preserve stability.