Learn More
Medchemexpress LLC 8-bromooct-1-yne | 81216-13-9 | MFCD20621358 | 189.09 g·mol⁻¹ | C8H13Br | 1 G
SDP
Catalog No. 5000285009
Encompass_Preferred
8-Bromooct-1-yne is a brominated terminal alkyne reagent used as a synthetic building block in organic synthesis and chemical biology research. It combines a terminal alkyne for click-type reactions with a bromide leaving group for nucleophilic substitution, enabling installation of alkynyl linkers and chain elaboration in probe and linker synthesis.
  • Terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Bromide functions as a good leaving group for nucleophilic substitution and chain modification.
  • Suitable as a building block for probes, tags, and linker or PROTAC synthesis.
  • Available in small laboratory quantities for research use.
  • Handle under inert atmosphere and store cold according to handling guidelines.
Catalog No. 50-002-85009 Supplier Medchemexpress LLC Supplier No. HYW5919701G
8-Bromooct-1-yne is a brominated terminal alkyne reagent used as a synthetic building block in organic synthesis and chemical biology research. It combines a terminal alkyne for click-type reactions with a bromide leaving group for nucleophilic substitution, enabling installation of alkynyl linkers and chain elaboration in probe and linker synthesis.
  • Terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Bromide functions as a good leaving group for nucleophilic substitution and chain modification.
  • Suitable as a building block for probes, tags, and linker or PROTAC synthesis.
  • Available in small laboratory quantities for research use.
  • Handle under inert atmosphere and store cold according to handling guidelines.