Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-bromooct-1-yne | 81216-13-9 | MFCD20621358 | 189.09 g/mol | C8H13Br | 100 MG
SDP

Catalog No. 5000284527
Encompass_Preferred

8-Bromooct-1-yne is a brominated terminal alkyne reagent used in organic synthesis and copper-catalyzed click chemistry. It offers both a reactive alkyne for azide coupling and a bromide leaving group for substitution, supplied in small research-scale quantities and requiring inert storage.

  • Terminal alkyne reagent for organic synthesis and click chemistry.
  • Contains a bromide leaving group for nucleophilic substitution.
  • Molecular formula C8H13Br; molecular weight 189.09 g/mol.
  • Suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Available in small research-scale pack sizes for laboratory use.
  • Store under nitrogen at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-84527 Supplier Medchemexpress LLC Supplier No. HYW591970100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Bromooct-1-yne is a brominated terminal alkyne reagent used in organic synthesis and copper-catalyzed click chemistry. It offers both a reactive alkyne for azide coupling and a bromide leaving group for substitution, supplied in small research-scale quantities and requiring inert storage.

  • Terminal alkyne reagent for organic synthesis and click chemistry.
  • Contains a bromide leaving group for nucleophilic substitution.
  • Molecular formula C8H13Br; molecular weight 189.09 g/mol.
  • Suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Available in small research-scale pack sizes for laboratory use.
  • Store under nitrogen at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.