Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-bromooct-1-yne | 81216-13-9 | MFCD20621358 | 189.09 g/mol | C8H13Br | 250 MG
SDP

Catalog No. 5000284525
Encompass_Preferred

8-Bromooct-1-yne is a terminal alkyne bearing a bromide substituent used as a synthetic building block for organic synthesis and chemical biology. It participates in alkyne-based coupling and substitution reactions, enabling installation of functional groups or linkers in small-molecule synthesis and bioconjugation.

  • Terminal alkyne with a reactive bromide group.
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (click chemistry) and cross-coupling reactions.
  • Useful as an alkylating building block for functionalization and chain extension.
  • Provided as a 250 mg research reagent; store under inert atmosphere at low temperature for stability.
  • Molecular formula C8H13Br; molecular weight 189.09 g/mol.
  • Purity not specified on the manufacturer product page.

Catalog No. 50-002-84525 Supplier Medchemexpress LLC Supplier No. HYW591970250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Bromooct-1-yne is a terminal alkyne bearing a bromide substituent used as a synthetic building block for organic synthesis and chemical biology. It participates in alkyne-based coupling and substitution reactions, enabling installation of functional groups or linkers in small-molecule synthesis and bioconjugation.

  • Terminal alkyne with a reactive bromide group.
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (click chemistry) and cross-coupling reactions.
  • Useful as an alkylating building block for functionalization and chain extension.
  • Provided as a 250 mg research reagent; store under inert atmosphere at low temperature for stability.
  • Molecular formula C8H13Br; molecular weight 189.09 g/mol.
  • Purity not specified on the manufacturer product page.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.