Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-bromotheophylline | 10381-75-6 | 99.3% | 259.06 g/mol | C7H7BrN4O2 | 250 MG
SDP

Catalog No. 5000208693
Encompass_Preferred

8-Bromotheophylline is a brominated theophylline derivative supplied for research and analytical applications and as a drug intermediate used in the synthesis of oxazolo[2,3-f]purinediones evaluated for adenosine receptor affinity.

  • High purity of 99.3%.
  • White to off-white solid appearance.
  • Chemical formula C7H7BrN4O2; molecular weight 259.06 g/mol.
  • Soluble in DMSO at 25 mg/mL; ultrasonic treatment recommended.
  • Storage: store at 4°C sealed, away from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
  • Available in solid pack sizes including 250 mg.

Catalog No. 50-002-08693 Supplier Medchemexpress LLC Supplier No. 250MGHYW088152
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Bromotheophylline is a brominated theophylline derivative supplied for research and analytical applications and as a drug intermediate used in the synthesis of oxazolo[2,3-f]purinediones evaluated for adenosine receptor affinity.

  • High purity of 99.3%.
  • White to off-white solid appearance.
  • Chemical formula C7H7BrN4O2; molecular weight 259.06 g/mol.
  • Soluble in DMSO at 25 mg/mL; ultrasonic treatment recommended.
  • Storage: store at 4°C sealed, away from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
  • Available in solid pack sizes including 250 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.