Learn More
Cayman Chemical 8-butylthio NAD sodium salt
SDP
Catalog No. 502917226
Encompass_Preferred
A derivative of NAD+; has been used in the screening of analog-sensitive PARP1 gatekeeper mutations
Catalog No. 50-291-7226 Supplier Cayman Chemical Supplier No. 38476800 UG
A derivative of NAD+; has been used in the screening of analog-sensitive PARP1 gatekeeper mutations