Learn More
MSC 8 CAL FACE SHIELD KIT
Catalog No. 5000803836
Encompass
This face shield and headgear combination provides personal protection, featuring a propionate window and orange headgear. It measures 10 in. high and 20 in. wide.
  • Face shield and headgear combination
  • Propionate window material
  • Orange headgear
  • Anti-fog window coating
  • Ratchet headgear adjustment
  • Nylon headgear material
  • Green window color
  • Window thickness 0.0600 in.
  • Window height 10 in.
  • Window width 20 in.
  • Lens shade 8
  • Not vented
  • Hard hat ANSI Type I
  • Hard hat class rating E
Catalog No. 50-008-03836 Supplier MSC Supplier No. 92258466
This face shield and headgear combination provides personal protection, featuring a propionate window and orange headgear. It measures 10 in. high and 20 in. wide.
  • Face shield and headgear combination
  • Propionate window material
  • Orange headgear
  • Anti-fog window coating
  • Ratchet headgear adjustment
  • Nylon headgear material
  • Green window color
  • Window thickness 0.0600 in.
  • Window height 10 in.
  • Window width 20 in.
  • Lens shade 8
  • Not vented
  • Hard hat ANSI Type I
  • Hard hat class rating E