Learn More
National Safety Apparel™ 8 Cal UltraSoft Arc Flash Bib Overalls
SDP
Catalog No. 17001340 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-340 5X-Large
17-001-333 Small
17-001-334 Medium
17-001-335 Large
17-001-336 X-Large
17-001-337 2X-Large
17-001-338 3X-Large
17-001-339 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-340 Supplier National Safety Apparel™ Supplier No. C45UJ5X32
Westex™ Ultrasoft is the ideal choice for superior comfort, improved safety, and above average durability.
  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 7 oz. Westex™ UltraSoft™
  • FR fabric will not melt, drip or ignite
  • Inherently flame resistant protection will not wash/wear out
  • Quick disconnect and easy adjustable shoulder straps for quick entry
  • FR hook and loop calf closure for easy boot entry
  • FR hook and loop fly closure
  • Made in the USA
Specifications
Closure Type Buckle
Color Navy
Material Cotton, Nylon
Seam Type Stitched
Size 5X-Large
Suspender Adjustable
Arc Flash Rating 8 cal/cm²
Waist Style Standard
Resistance Flame
Certifications/Compliance CAT 2, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Disposable Reusable