Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 8 cal Coverall
SDP

Catalog No. 17001445 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-445 Small
17-001-446 Medium
17-001-447 Large
17-001-448 X-Large
17-001-449 2X-Large
17-001-450 3X-Large
17-001-451 4X-Large
17-001-452 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-445 Supplier National Safety Apparel™ Supplier No. C88UJSM32
Add to Cart
Add to Cart

Coveralls with comfortable, sewn-in elastic waist

  • Comfortable, sewn-in elastic waist
  • Two-way covered zipper down front for easy entry
  • Breast pockets with flaps and hidden snap closures
  • Two large rear pockets
  • Two front pockets with pass-through slashes for access to pants pockets
  • Leg openings made to fit over work boots
  • All metal covered to ensure no contact with skin
  • Action back allows for full range of motion

Specifications

Product Type Coveralls
Size Small
Gender Unisex
Material Cotton, Nylon
Color Navy
Disposable Reusable
Waist Style Elastic (Stitched-in)
Ankle Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Covered Zipper
Seam Type Stitched
Sterility Non-sterile
Protection Class CAT 2
Resistance Flame
Arc Flash Rating 8 cal/cm²
Certifications/Compliance NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Suspender None
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Style Coveralls
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.