Learn More
National Safety Apparel™ 8 Cal UltraSoft FR Overpants
SDP
Catalog No. 170012301 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2301 5X-Large
17-001-2294 Small
17-001-2295 Medium
17-001-2296 Large
17-001-2297 X-Large
17-001-2298 2X-Large
17-001-2299 3X-Large
17-001-2300 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-2301 Supplier National Safety Apparel™ Supplier No. TSPUJ5X
Overpants designed for comfort and flexibility
  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 7 oz. Westex™ UltraSoft™
  • Designed for comfort and flexibility
  • Excellent layering piece for added protection
  • FR hook and loop leg closures
  • Generous cut allows for unrestricted movement and comfort
  • FR fabric will not melt, drip or ignite
  • Home laundry
  • No pockets
  • Made in the USA
Specifications
Size 5X-Large
Gender Unisex
Material Cotton, Nylon
Color Navy
Ankle Style Open
Seam Type Stitched
Closure Type Zipper
Disposable Reusable
Certifications/Compliance CAT 2, NFPA 70 E, NFPA, 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Sterility Non-sterile