Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 8 Channel, 96 Well, Standard, 19 Gauge, SS Tubes, 14mm Long, 9mm Spacing, Polycarbonate
Catalog No. NC9172325
Encompass
ASPIRATION MANIFOLD, 8 Channel, 96 Well, Standard Plate, 19 Gauge, Stainless Steel Tubes, 14mm Long, 9mm Center Spacing, Polycarbonate
Catalog No. NC9172325 Supplier V&P Scientific Inc Supplier No. VP 180
ASPIRATION MANIFOLD, 8 Channel, 96 Well, Standard Plate, 19 Gauge, Stainless Steel Tubes, 14mm Long, 9mm Center Spacing, Polycarbonate