Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 8 Channel, 96 Well, Standard, 19 Gauge, SS Tubes, 14mm Long, 9mm Spacing, Polypropylene
Catalog No. NC9024821
Encompass
ASPIRATION MANIFOLD, 8 Channel, 96 Well, Standard Plate, 19 Gauge, Stainless Steel Tubes, 14mm Long, 9mm Center Spacing, Polypropylene, Female Luer, Solvent Resistant
Catalog No. NC9024821 Supplier V&P Scientific Inc Supplier No. VP 180P
ASPIRATION MANIFOLD, 8 Channel, 96 Well, Standard Plate, 19 Gauge, Stainless Steel Tubes, 14mm Long, 9mm Center Spacing, Polypropylene, Female Luer, Solvent Resistant