Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 8 Channel, 48 Well, Standard, 19 Gauge, SS Tubes, 19mm Long, 13.4mm Spacing, Polycarbonate
Catalog No. NC9519328
Encompass
ASPIRATION MANIFOLD, 8 Channel, 48 Well, Standard Plate, 19 Gauge, Stainless Steel Tubes, 19mm Long, 13.4mm Center Spacing, Polycarbonate, for Costar 3548 Plates
Catalog No. NC9519328 Supplier V&P Scientific Inc Supplier No. VP 188
ASPIRATION MANIFOLD, 8 Channel, 48 Well, Standard Plate, 19 Gauge, Stainless Steel Tubes, 19mm Long, 13.4mm Center Spacing, Polycarbonate, for Costar 3548 Plates