Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 8 Channel, 48 Well, Deep Well, 19 Gauge, SS Tubes, 32mm Long, 13.4mm Spacing, Polycarbonate
Catalog No. NC0301210
Encompass
ASPIRATION MANIFOLD, 8 Channel, 48 Well, Deep Well Plate, 19 Gauge, Stainless Steel Tubes, 32mm Long, 13.4mm Center Spacing, Polycarbonate, for Costar 3548 Plates
Catalog No. NC0301210 Supplier V&P Scientific Inc Supplier No. VP 188B
ASPIRATION MANIFOLD, 8 Channel, 48 Well, Deep Well Plate, 19 Gauge, Stainless Steel Tubes, 32mm Long, 13.4mm Center Spacing, Polycarbonate, for Costar 3548 Plates