Learn More
Arcticwhite LLC 8 Row Partitioned Reservoir, Pyramid Bottom, Non-sterilized, 8 Rows, Partitioned, Polypropylene (PP), 32 ML/Row, 25 Reagent Reservoirs per Case
SDP
Catalog No. NC1117232
Encompass
8 Row Partitioned Reservoir, Pyramid Bottom, Non-sterile, 8 Rows, Partitioned, Polypropylene (PP), 32 ML/Row, 25 Reagent Reservoirs per Case. The 8-row reagent reservoir has partitioned channels and a pyramid bottom. It suits automated liquid handlers and multichannel pipetting. Format - 8 rows, partitioned. Bottom - Pyramid. Capacity - 32 mL/Row. Material - Polypropylene (PP). Supplied by ArcticWhite as 25 reagent reservoirs per case.
Catalog No. NC1117232 Supplier Arcticwhite LLC Supplier No. AWLSS30020
8 Row Partitioned Reservoir, Pyramid Bottom, Non-sterile, 8 Rows, Partitioned, Polypropylene (PP), 32 ML/Row, 25 Reagent Reservoirs per Case. The 8-row reagent reservoir has partitioned channels and a pyramid bottom. It suits automated liquid handlers and multichannel pipetting. Format - 8 rows, partitioned. Bottom - Pyramid. Capacity - 32 mL/Row. Material - Polypropylene (PP). Supplied by ArcticWhite as 25 reagent reservoirs per case.