Learn More
ANALYTIK JENA US LLC Analytik Jena 8-Channel Adapter, Multi-Channel Pipetting Adapter, Positive Air Displacement, 1 unit
Catalog No. NC3734416
Encompass
The Analytik Jena 8-Channel Adapter is designed for the CyBio FeliX liquid handling system, enabling efficient multi-channel pipetting. It enhances workflow automation, supports various plate formats (96-well and 384-well), and utilizes positive air displacement technology for precise liquid transfers. Its modular and space-saving design facilitates diverse applications, including sample preparation and advanced sequencing tasks, making it a versatile solution for high-throughput environments.
Catalog No. NC3734416 Supplier ANALYTIK JENA US LLC Supplier No. OL331714330
Add to Cart
Add to Cart
Request a Quote
The Analytik Jena 8-Channel Adapter is designed for the CyBio FeliX liquid handling system, enabling efficient multi-channel pipetting. It enhances workflow automation, supports various plate formats (96-well and 384-well), and utilizes positive air displacement technology for precise liquid transfers. Its modular and space-saving design facilitates diverse applications, including sample preparation and advanced sequencing tasks, making it a versatile solution for high-throughput environments.