Learn More
Waters Corp Deepwell Microplate Domino, Machine-Readable, Andrew+ Compatible, 1 pk
Supplier:  Waters Corp 186009597
Encompass_Preferred
The Deepwell Microplate Domino is designed to hold Deepwell plates and reagent reservoirs, serving as a machine-readable consumable holder that interfaces with the Andrew+ pipetting robot for seamless automation of laboratory workflows. It features QR codes for labware information and ARUCO codes for spatial positioning.
  • Holds Deepwell plates and reagent reservoirs
  • Machine-readable consumable holders
  • Equipped with QR codes for labware information
  • Features ARUCO codes for spatial positioning
  • Designed to interface with the Andrew+ pipetting robot
  • Enables seamless automation of laboratory workflows
  • 1 unit per package
Catalog No. 50-226-9320