Learn More
Waters Corp Andrew Alliance Pipette, Single-channel, Electronic, 10 to 300 μL
Catalog No. 502269322 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Bluetooth electronic single-channel pipette provides accurate and precise pipetting for use with the Andrew+ pipetting robot, Pipette+ guided pipetting system, or manual pipetting. Its ergonomic design helps prevent repetitive strain injuries. Fully electronic operation reduces variability, and Bluetooth communication with Andrew+ or Pipette+, and OneLab enables remote programming and pipetting traceability.
  • Intuitive user interface.
  • Autoclavable lower parts (except for 1200 μL multi-channel lower parts).
  • Optiload for perfect tip sealing.
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing and more.
  • Safe-cone filters in all models >10 μL to prevent contamination.
This item is not returnable. View return policy
Catalog No. 50-226-9322 Supplier Waters Corp Supplier No. 186009606
This Bluetooth electronic single-channel pipette provides accurate and precise pipetting for use with the Andrew+ pipetting robot, Pipette+ guided pipetting system, or manual pipetting. Its ergonomic design helps prevent repetitive strain injuries. Fully electronic operation reduces variability, and Bluetooth communication with Andrew+ or Pipette+, and OneLab enables remote programming and pipetting traceability.
  • Intuitive user interface.
  • Autoclavable lower parts (except for 1200 μL multi-channel lower parts).
  • Optiload for perfect tip sealing.
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing and more.
  • Safe-cone filters in all models >10 μL to prevent contamination.