Learn More
Waters Corp Waters Corporation Electronic Pipette, 8-Channel, 0.2–10 μL
Catalog No. 502269328 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Bluetooth Electronic Pipette, compatible with Guided Pipetting Systems and Robots, connects to intuitive cloud-native software, offering sophistication and ergonomics for laboratory tasks.
  • Intuitive interface with hot key for quick program access and adjustment wheel for volume setting.
  • Available in English, Chinese, French, German, and Russian.
  • Ergonomic design with comfortable handle, finger hook, and electronic tip ejection.
  • Bluetooth connectivity for two-way software communication and remote pipette programming.
  • Electronic brake and piston control ensure accurate, user-independent results.
  • Optional Microwell Plate Tracker guides pipetting for 96- and 384-well plates.
  • Supports multiple pipetting modes: pipetting, mixing, counter, auto dispensing, multi dispensing, diluting, and titrate.
This item is not returnable. View return policy
Catalog No. 50-226-9328 Supplier Waters Corp Supplier No. 186009767
This Bluetooth Electronic Pipette, compatible with Guided Pipetting Systems and Robots, connects to intuitive cloud-native software, offering sophistication and ergonomics for laboratory tasks.
  • Intuitive interface with hot key for quick program access and adjustment wheel for volume setting.
  • Available in English, Chinese, French, German, and Russian.
  • Ergonomic design with comfortable handle, finger hook, and electronic tip ejection.
  • Bluetooth connectivity for two-way software communication and remote pipette programming.
  • Electronic brake and piston control ensure accurate, user-independent results.
  • Optional Microwell Plate Tracker guides pipetting for 96- and 384-well plates.
  • Supports multiple pipetting modes: pipetting, mixing, counter, auto dispensing, multi dispensing, diluting, and titrate.