Learn More
New Era Pump Systems Inc 8 CHANNEL PUMP
Catalog No. NC1329132
Encompass_Preferred
Holds 2, 4, 6, or 8 syringes of up to 10 mL each. Infusion rates from 0.452 ¿L/hr (1 mL syringe) to 415.9 mL/hr (10 mL syringe). Easy-to-use keypad interface. Higher flow rate models available. Includes all the advanced functions and programming features of the NE-1000 series. Disposable, Glass, Stainless Steel Syringes and Plumbing Supplies are sold separately.
Catalog No. NC1329132 Supplier New Era Pump Systems Inc Supplier No. NE1800
Holds 2, 4, 6, or 8 syringes of up to 10 mL each. Infusion rates from 0.452 ¿L/hr (1 mL syringe) to 415.9 mL/hr (10 mL syringe). Easy-to-use keypad interface. Higher flow rate models available. Includes all the advanced functions and programming features of the NE-1000 series. Disposable, Glass, Stainless Steel Syringes and Plumbing Supplies are sold separately.