Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(8-Chloro-1-naphthylthio)acetic Acid 95.0+%, TCI America™
SDP

Catalog No. C20525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20525G Supplier TCI America Supplier No. C20525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 129-94-2
Molecular Formula C12H8ClO2S
Molecular Weight (g/mol) 251.70
MDL Number MFCD00043901
InChI Key WPNAGQJVWAFQJV-UHFFFAOYSA-M
Synonym 8-chloro-1-naphthylthio acetic acid, 2-8-chloro-1-naphthylthio acetic acid, 2-8-chloronaphthalen-1-yl sulfanyl acetic acid, 2-8-chloronaphthalen-1-yl thio acetic acid, 8-chloronaphthalen-1-yl sulfanyl acetic acid, 2-8-chloro-1-naphthyl thio acetic acid, maybridge1_001748, 8-chloro-1-naphthyl sulfanyl acetic acid, acmc-1bugo, 2-8-chloronaphthylthio acetic acid
PubChem CID 67219
IUPAC Name 2-[(8-chloronaphthalen-1-yl)sulfanyl]acetate
SMILES [O-]C(=O)CSC1=C2C(Cl)=CC=CC2=CC=C1

Specifications

Melting Point 157°C
Color Yellow
Quantity 5 g
Formula Weight 252.71
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (8-Chloro-1-naphthylthio)acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.