Learn More
Medchemexpress LLC 8-chloro-11-(4-ethylpiperazin-1-yl)-1-fluoro-5H-dibenzo[b,e]diazepine | 2369979-67-7 | 99.5% | 358.84 g/mol | C19H20ClFN4 | 5 MG
SDP
Catalog No. 5000262836
Encompass_Preferred
A brain-penetrant DREADD agonist research compound with low-nanomolar activity at engineered muscarinic receptors. It demonstrates selective displacement of radioligand binding in brain tissue and is supplied as a high-purity solid or as ready-to-use DMSO solutions for research applications. For research use only; not for human or clinical use.
  • Potent agonist at hM3Dq (EC50 ≈ 5 nM) and hM4Di (EC50 ≈ 0.5 nM).
  • Brain-penetrant with selective displacement of [3H]clozapine binding in brain tissue.
  • High purity solid (≈99.5%) and available as 10 mM DMSO solutions for convenience.
  • Suitable for in vivo and in vitro DREADD activation and imaging studies.
  • Provided for research use only; not intended for human or clinical applications.
Catalog No. 50-002-62836 Supplier Medchemexpress LLC Supplier No. HY1318915MG
Add to Cart
Add to Cart
A brain-penetrant DREADD agonist research compound with low-nanomolar activity at engineered muscarinic receptors. It demonstrates selective displacement of radioligand binding in brain tissue and is supplied as a high-purity solid or as ready-to-use DMSO solutions for research applications. For research use only; not for human or clinical use.
  • Potent agonist at hM3Dq (EC50 ≈ 5 nM) and hM4Di (EC50 ≈ 0.5 nM).
  • Brain-penetrant with selective displacement of [3H]clozapine binding in brain tissue.
  • High purity solid (≈99.5%) and available as 10 mM DMSO solutions for convenience.
  • Suitable for in vivo and in vitro DREADD activation and imaging studies.
  • Provided for research use only; not intended for human or clinical applications.