Learn More
Medchemexpress LLC 8-chloro-11-(4-ethylpiperazin-1-yl)-4-fluoro-5H-dibenzo[b,e]diazepine | 2369979-68-8 | 100.0% | C19H20ClFN4 | 10MG
SDP
Catalog No. 5000261244
Encompass_Preferred
JHU37160 is a potent, brain-penetrant small-molecule agonist for Designer Receptors Exclusively Activated by Designer Drugs (DREADDs), active at hM3Dq and hM4Di. It is characterized by CAS 2369979-68-8, molecular formula C19H20ClFN4, and molecular weight 358.84, and is provided for research applications.
  • Potent agonist for hM3Dq and hM4Di
  • Brain-penetrant small molecule
  • High reported purity (99.98%)
  • Available as solid and solution formulations
  • Defined storage stability in powder and solvent
Catalog No. 50-002-61244 Supplier Medchemexpress LLC Supplier No. HY13188110MG
JHU37160 is a potent, brain-penetrant small-molecule agonist for Designer Receptors Exclusively Activated by Designer Drugs (DREADDs), active at hM3Dq and hM4Di. It is characterized by CAS 2369979-68-8, molecular formula C19H20ClFN4, and molecular weight 358.84, and is provided for research applications.
  • Potent agonist for hM3Dq and hM4Di
  • Brain-penetrant small molecule
  • High reported purity (99.98%)
  • Available as solid and solution formulations
  • Defined storage stability in powder and solvent