Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-chloro-2-((2,4-dichlorophenyl)amino)-3-(3-methylbutanoyl)-5-nitroquinolin-4(1H)-one | 1361227-90-8 | 99.9% | 468.72 | C20H16Cl3N3O4 | 25MG
SDP

Catalog No. 5000239229
Encompass_Preferred

JH-RE-06 is a small-molecule inhibitor that disrupts the REV1-REV7 interface, preventing recruitment of mutagenic POLζ and enhancing tumor cell sensitivity to DNA-damaging agents. It is provided for research use with reported activity data and analytical documentation available.

  • Targets the REV1-REV7 interface to disrupt mutagenic translesion synthesis.
  • Reported IC50 0.78 μM and Kd 0.42 μM for activity characterization.
  • Chemical formula C20H16Cl3N3O4 and molecular weight 468.72 g·mol⁻¹.
  • High purity: 99.87% (reported).
  • Available in multiple solid sizes including 25 mg and as 10 mM solution in DMSO.
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Analytical documentation includes datasheet, COA, SDS, and LCMS.

Catalog No. 50-002-39229 Supplier Medchemexpress LLC Supplier No. HY12621425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JH-RE-06 is a small-molecule inhibitor that disrupts the REV1-REV7 interface, preventing recruitment of mutagenic POLζ and enhancing tumor cell sensitivity to DNA-damaging agents. It is provided for research use with reported activity data and analytical documentation available.

  • Targets the REV1-REV7 interface to disrupt mutagenic translesion synthesis.
  • Reported IC50 0.78 μM and Kd 0.42 μM for activity characterization.
  • Chemical formula C20H16Cl3N3O4 and molecular weight 468.72 g·mol⁻¹.
  • High purity: 99.87% (reported).
  • Available in multiple solid sizes including 25 mg and as 10 mM solution in DMSO.
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Analytical documentation includes datasheet, COA, SDS, and LCMS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.