Learn More
Cayman Chemical 8-chloro Caffeine, Purity: ≥98%, CAS: 4921-49-7, 250 mg
SDP
Catalog No. 502912434
Encompass_Preferred
A derivative of caffeine; binds to adenosine receptors (apparent Ki = 30 µM); potentiates UV-induced chromosomal aberrations in Cl-I Chinese hamster embryonic lung cells at 250 µM; has been used in the synthesis of MLKL inhibitors with necroptosis-inhibiting activity
Catalog No. 50-291-2434 Supplier Cayman Chemical Supplier No. 35817250 MG
A derivative of caffeine; binds to adenosine receptors (apparent Ki = 30 µM); potentiates UV-induced chromosomal aberrations in Cl-I Chinese hamster embryonic lung cells at 250 µM; has been used in the synthesis of MLKL inhibitors with necroptosis-inhibiting activity