Learn More
Medchemexpress LLC 8-chloro caffeine | 4921-49-7 | MFCD00047246 | 99.4% | 228.64 | C8H9ClN4O2 | 1 G
SDP
Catalog No. 5000282207
Encompass_Preferred
8-Chloro caffeine is a halogenated caffeine analogue used as a research reagent that binds adenosine receptors (reported Ki ≈ 30 μM). Supplied as a solid powder, it is intended for in vitro pharmacology and receptor binding studies; handle with appropriate laboratory safety procedures and follow storage recommendations.
Catalog No. 50-002-82207 Supplier Medchemexpress LLC Supplier No. HYW1783271G
8-Chloro caffeine is a halogenated caffeine analogue used as a research reagent that binds adenosine receptors (reported Ki ≈ 30 μM). Supplied as a solid powder, it is intended for in vitro pharmacology and receptor binding studies; handle with appropriate laboratory safety procedures and follow storage recommendations.