Learn More
Medchemexpress LLC 8-chloro caffeine | 4921-49-7 | MFCD00047246 | 99.4% | 228.64 g·mol⁻¹ | C8H9ClN4O2 | 100 MG
SDP
Catalog No. 5000283228
Encompass_Preferred
8-Chloro caffeine is a chlorinated derivative of the methylxanthine alkaloid caffeine that binds adenosine receptors (reported Ki = 30 μM). It is supplied as a white to off-white solid for laboratory research and includes documented physicochemical and storage recommendations.
  • Binds adenosine receptors (reported Ki = 30 μM).
  • Derivative of caffeine useful for receptor-binding and pharmacology studies.
  • Molecular formula C8H9ClN4O2; molecular weight 228.64 g·mol⁻¹.
  • High purity (approximately 99.4%).
  • Solid, white to off-white appearance for easy handling.
  • Store powder at -20°C for long-term stability; store in solvent at -80°C for up to 6 months.
Catalog No. 50-002-83228 Supplier Medchemexpress LLC Supplier No. HYW178327100MG
8-Chloro caffeine is a chlorinated derivative of the methylxanthine alkaloid caffeine that binds adenosine receptors (reported Ki = 30 μM). It is supplied as a white to off-white solid for laboratory research and includes documented physicochemical and storage recommendations.
  • Binds adenosine receptors (reported Ki = 30 μM).
  • Derivative of caffeine useful for receptor-binding and pharmacology studies.
  • Molecular formula C8H9ClN4O2; molecular weight 228.64 g·mol⁻¹.
  • High purity (approximately 99.4%).
  • Solid, white to off-white appearance for easy handling.
  • Store powder at -20°C for long-term stability; store in solvent at -80°C for up to 6 months.