Learn More
Medchemexpress LLC 8-chloro caffeine | 4921-49-7 | MFCD00047246 | 99.4% | 228.64 g/mol | C8H9ClN4O2 | 25 MG
SDP
Catalog No. 5000281554
Encompass_Preferred
8-Chloro caffeine is a chlorinated analog of caffeine used as a research tool to probe adenosine receptor activity. It is supplied as a solid powder for laboratory use, with reported adenosine receptor binding (Ki ≈ 30 μM), high purity, and a molecular weight of 228.64 g/mol. Typical uses include receptor screening and biochemical assays.
  • Adenosine receptor binder (Ki ≈ 30 μM).
  • High purity suitable for research applications (≈99.4%).
  • Molecular formula C8H9ClN4O2; molecular weight 228.64 g/mol.
  • Solid powder format for convenient storage and handling.
  • Small pack sizes ideal for screening and assay development.
Catalog No. 50-002-81554 Supplier Medchemexpress LLC Supplier No. HYW17832725MG
8-Chloro caffeine is a chlorinated analog of caffeine used as a research tool to probe adenosine receptor activity. It is supplied as a solid powder for laboratory use, with reported adenosine receptor binding (Ki ≈ 30 μM), high purity, and a molecular weight of 228.64 g/mol. Typical uses include receptor screening and biochemical assays.
  • Adenosine receptor binder (Ki ≈ 30 μM).
  • High purity suitable for research applications (≈99.4%).
  • Molecular formula C8H9ClN4O2; molecular weight 228.64 g/mol.
  • Solid powder format for convenient storage and handling.
  • Small pack sizes ideal for screening and assay development.