Learn More
Medchemexpress LLC 8-chloro caffeine | 4921-49-7 | MFCD00047246 | 99.4% | 228.64 g·mol⁻¹ | C8H9ClN4O2 | 50 MG
SDP
Catalog No. 5000281898
Encompass_Preferred
8-Chloro caffeine is a halogenated caffeine analogue used in research as an adenosine receptor ligand. It has been reported to bind adenosine receptors (apparent Ki ≈ 30 μM) and is supplied as a solid for laboratory use. The compound's CAS number is 4921-49-7 and molecular weight is 228.64 g·mol⁻¹.
Catalog No. 50-002-81898 Supplier Medchemexpress LLC Supplier No. HYW17832750MG
8-Chloro caffeine is a halogenated caffeine analogue used in research as an adenosine receptor ligand. It has been reported to bind adenosine receptors (apparent Ki ≈ 30 μM) and is supplied as a solid for laboratory use. The compound's CAS number is 4921-49-7 and molecular weight is 228.64 g·mol⁻¹.