Learn More
Cayman Chemical 8-Chloroadenosine, Purity: ≥98%, CAS: 34408-14-5, 10 mg
SDP
Catalog No. 502911402
Encompass_Preferred
A nucleoside analog and an active metabolite of 8-chloro cAMP; inhibits RNA but not DNA synthesis in cells at 10 µM; is cytotoxic to patient-derived multiple myeloma cells; reduces tumor growth by 50% in an HCT116 colorectal cancer mouse xenograft model at 50 mg/kg
Catalog No. 50-291-1402 Supplier Cayman Chemical Supplier No. 3576610 MG
A nucleoside analog and an active metabolite of 8-chloro cAMP; inhibits RNA but not DNA synthesis in cells at 10 µM; is cytotoxic to patient-derived multiple myeloma cells; reduces tumor growth by 50% in an HCT116 colorectal cancer mouse xenograft model at 50 mg/kg