Learn More
Enzo Life Sciences 8-Chloroadenosine (5mg). CAS: 34408-14-5
SDP
Supplier:  Enzo Life Sciences ALX480028M005
Encompass_Preferred
Cytotoxic metabolite of 8-Cl-cAMP (ALX-480-029). Purity: ≥98% (HPLC). Appearance: Crystalline solid. Solubility: Soluble water (100mM at 50°C). Long Term Storage: +4°C.
Catalog No. 50-200-9309