Learn More
AdipoGen 8-Chlorotheophylline
SDP
Supplier:  AdipoGen CDXC0153G100
Encompass_Preferred
Chemical. CAS 85-18-7. Formula C7H7ClN4O2. MW 214.61. Synthetic Stimulant slightly ionotropic drug of the xanthine chemical class, with physiological effects similar to caffeine. It is combined with pharmaceutical drugs to form stable salts.
Catalog No. 50-256-1452