Learn More
CELLTREAT Scientific Products 8 Chamber Cell Culture Slide, No Marking Spot, Glass, Sterile
SDP
Catalog No. NC3412498
Encompass_Preferred
Grow cells directly on the slide without any need for future transfer; ideal for growing and viewing cells under a microscope on standard 25mm x 75mm slideUniform hydrophilic surface ideal for consistent cell attachmentSlides are labeled with numeric markingsPacking trays can be used as incubation racks in CO2 incubatorsAdhesive freeReusable slide tray available (PN 229167)Integrated gasket prevents cross-contamination between chambersInert hydrophobic green border printed on slide to define cell culture areaChamber holder features easy flip locks assembly and disassembly; no separation tool requiredNew ""No Marking Spot"" slide is ideal for applications involving solvents"
Catalog No. NC3412498 Supplier CELLTREAT Scientific Products Supplier No. 229169
Grow cells directly on the slide without any need for future transfer; ideal for growing and viewing cells under a microscope on standard 25mm x 75mm slideUniform hydrophilic surface ideal for consistent cell attachmentSlides are labeled with numeric markingsPacking trays can be used as incubation racks in CO2 incubatorsAdhesive freeReusable slide tray available (PN 229167)Integrated gasket prevents cross-contamination between chambersInert hydrophobic green border printed on slide to define cell culture areaChamber holder features easy flip locks assembly and disassembly; no separation tool requiredNew ""No Marking Spot"" slide is ideal for applications involving solvents"