Learn More
Enzo Life Sciences 8-CPT-cAMP (100mg). CAS: 93882-12-3
SDP
Catalog No. 502010443
Encompass_Preferred
A lipophilic, cell-permeable cAMP analog which activates cAMP- and cGMP-dependent protein kinase.  More potent than cAMP and more resistant to phosphodiesterasese, with good selectivity for site B of PKA. For specific activation of the cAMP pathway the highly membrane permeable Sp-5,6-DCI-cBIMPS (BML-CN135). Purity: ≥96% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0443 Supplier Enzo Life Sciences Supplier No. BMLCN1300100
A lipophilic, cell-permeable cAMP analog which activates cAMP- and cGMP-dependent protein kinase.  More potent than cAMP and more resistant to phosphodiesterasese, with good selectivity for site B of PKA. For specific activation of the cAMP pathway the highly membrane permeable Sp-5,6-DCI-cBIMPS (BML-CN135). Purity: ≥96% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.